(S)-(-)-6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D4375-1G
D4375-5G
TCD4375-1GEA
50.56
CAD
TCD4375-1G
TCD4375-5G
(S)-(-)-6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride ≥98.0% (by HPLC, titration analysis)
(S)-(-)-6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride
CAS Number: 82586-62-7
MDL Number: MFCD01861226
Molecular Formula: C12H15NO4
Molecular Weight: 273.71
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 272
Formula:
C₁₂H₁₅NO₄ Melting Pt: 272 °C |
MDL Number:
MFCD01861226 CAS Number: 82586-62-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...