Dibenzothiophene-2-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Dibenzo[b,d]thiophen-2-ylboronic acid

D4373-1G D4373-5G
TCD4373-1GEA 88.48 CAD
TCD4373-1G TCD4373-5G
Dibenzothiophene-2-boronic acid (contains varying amounts of Anhydride)
Dibenzothiophene-2-boronic acid

CAS Number: 668983-97-9
MDL Number: MFCD01318982
Molecular Formula: C12H9BO2S
Molecular Weight: 228.07
Form: Crystal
Color: White

Formula: C₁₂H₉BO₂S
MDL Number: MFCD01318982
CAS Number: 668983-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36a792661874d5ead4f60da820b6f3a83f7809ef&vid=36a792661874d5ead4f60da820b6f3a83f7809ef&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR