1,2-Bis(1-naphthyl)ethane ≥94.0%

Supplier: TCI America

Synonyms: 1,2-Di(naphthalen-1-yl)ethane

D4371-1G D4371-5G
TCD4371-1GEA 153.73 CAD
TCD4371-1G TCD4371-5G
1,2-Bis(1-naphthyl)ethane ≥94.0%
1,2-Bis(1-naphthyl)ethane

CAS Number: 15374-45-5
MDL Number: MFCD00037736
Molecular Formula: C22H18
Molecular Weight: 282.39
Purity/Analysis Method: >94.0% (GC)
Form: Crystal
Melting point (°C): 161
Lambda max.: 430 nm (Benzene)


Other Product Names / Synonyms: 1,2-Bis(1-naphthyl)ethane

Formula: C₂₂H₁₈
MW: 282.3783 g/mol
Melting Pt: 161 °C
MDL Number: MFCD00037736
CAS Number: 15374-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5a60b7cc3e8c61b113017610edd8f86243d744a&vid=c5a60b7cc3e8c61b113017610edd8f86243d744a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR