2,7-Dibromophenanthrene-9,10-dione ≥97.0% (by GC)

Supplier: TCI America

D4363-1G D4363-5G
TCD4363-1GEA 185.18 CAD
TCD4363-1G TCD4363-5G
2,7-Dibromophenanthrene-9,10-dione ≥97.0% (by GC)
2,7-Dibromophenanthrene-9,10-dione

CAS Number: 84405-44-7
MDL Number: MFCD03931078
Molecular Formula: C14H6Br2O2
Molecular Weight: 366.01
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 331

Formula: C₁₄H₆Br₂O₂
Melting Pt: 331 °C
MDL Number: MFCD03931078
CAS Number: 84405-44-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b865d9003110e8525c31509423894a929d6e7522&vid=b865d9003110e8525c31509423894a929d6e7522&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR