(3-(Diethylamino)propyl)trimethoxysilane ≥97.0%

Supplier: TCI America

Synonyms: Trimethoxy(3-(diethylamino)propyl)silane, N,N-Diethyl-3-(trimethoxysilyl)propan-1-amine, (3-Diethylaminopropyl)trimethoxysilane, [3-(Diethylamino)propyl]trimethoxysilane, N,N-Diethyl-3-(trimethoxysilyl)propylamine

D4362-25ML D4362-5ML
TCD4362-25MLEA 194.28 CAD
TCD4362-25ML TCD4362-5ML
(3-(Diethylamino)propyl)trimethoxysilane ≥97.0%
(3-(Diethylamino)propyl)trimethoxysilane

CAS Number: 41051-80-3
MDL Number: MFCD00039882
Molecular Formula: C10H25NO3Si
Molecular Weight: 235.40
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 100
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: N,N-Diethyl-3-(trimethoxysilyl)propylamine

Formula: (H₃CCH₂)₂NCH₂CH₂CH₂Si(OCH₃)₃
MW: 235.4
Boiling Pt: 120 °C
Density: 0.94 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00039882
CAS Number: 41051-80-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cac6649a286c757f52d8e3928fc59f5a4018af7&vid=9cac6649a286c757f52d8e3928fc59f5a4018af7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR