2,4-Dihydroxy-6-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-Methyl-2,4-pyridinediol, 4,6-Dihydroxy-2-picoline, 4-Hydroxy-6-methyl-2-pyridone, 4-Hydroxy-6-methylpyridin-2(1H)-one

D4361-1G D4361-5G
TCD4361-1GEA 69.52 CAD
TCD4361-1G TCD4361-5G
2,4-Dihydroxy-6-methylpyridine ≥98.0% (by GC, titration analysis)
2,4-Dihydroxy-6-methylpyridine

CAS Number: 3749-51-7
MDL Number: MFCD01860849
Molecular Formula: C6H7NO2
Molecular Weight: 125.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 330
Lambda max.: 282 nm (EtOH)


Other Product Names / Synonyms: 6-Methyl-2,4-pyridinediol, 4,6-Dihydroxy-2-picoline, 4-Hydroxy-6-methyl-2-pyridone

Formula: C₆H₇NO₂
MW: 125.13 g/mol
Melting Pt: 330 °C
MDL Number: MFCD01860849
CAS Number: 3749-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d874a727ecd4df49530ff6eade247a136d78f90&vid=6d874a727ecd4df49530ff6eade247a136d78f90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR