2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0% (by GC)

Supplier: TCI America

D4357-1G D4357-5G
TCD4357-1GEA 172.22 CAD
TCD4357-1G TCD4357-5G
2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0% (by GC)
2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

CAS Number: 185990-03-8
MDL Number: MFCD05664111
Molecular Formula: C14H23BO2Si
Molecular Weight: 262.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 110
Specific Gravity (20/20): 0.97


Product Categories: Synthetic Organic Chemistry, Boration
Other Product Names / Synonyms: (Dimethylphenylsilyl)boronic Acid Pinacol Ester

Formula: C₁₄H₂₃BO₂Si
Boiling Pt: 120 °C
Density: 0.97 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD05664111
CAS Number: 185990-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4d97ea704d22612f12b4ba98266dd5122ef34f2&vid=d4d97ea704d22612f12b4ba98266dd5122ef34f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR