1-Decyl-3-methylimidazoliumbis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1-Decyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide, 1-Decyl-3-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide
D4351-25G
D4351-5G
TCD4351-25GEA
737.23
CAD
TCD4351-25G
TCD4351-5G
1-Decyl-3-methylimidazoliumbis(trifluoromethanesulfonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Decyl-3-methylimidazoliumbis(trifluoromethanesulfonyl)imide
CAS Number: 433337-23-6
MDL Number: MFCD12022376
Molecular Formula: C16H27F6N3O4S2
Molecular Weight: 503.52
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Clear Liquid
Melting point (°C): -19
Specific Gravity (20/20): 1.29
Formula:
C₁₆H₂₇F₆N₃O₄S₂ MW: 503.52 g/mol Melting Pt: –19 °C Density: 1.29 (20 °C) |
MDL Number:
MFCD12022376 CAS Number: 433337-23-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...