2,6-Dimethylphenoxyacetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(2,6-Dimethylphenoxy)acetic acid
D4345-1G
D4345-5G
TCD4345-1GEA
85.32
CAD
TCD4345-1G
TCD4345-5G
2,6-Dimethylphenoxyacetic acid ≥98.0% (by GC, titration analysis)
2,6-Dimethylphenoxyacetic acid
CAS Number: 13335-71-2
MDL Number: MFCD00156912
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 139
Formula:
C₁₀H₁₂O₃ MW: 180.20 g/mol Melting Pt: 139 °C |
MDL Number:
MFCD00156912 CAS Number: 13335-71-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...