5'-Deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoropyrimidin-2(1H)-one

D4342-1G D4342-5G
TCD4342-1GEA 66.36 CAD
TCD4342-1G TCD4342-5G
5'-Deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)
5'-Deoxy-5-fluorocytidine

CAS Number: 66335-38-4
MDL Number: MFCD07369278
Molecular Formula: C9H12FN3O4
Molecular Weight: 245.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 204

Formula: C₉H₁₂FN₃O₄
MW: 245.21 g/mol
Melting Pt: 204 °C
MDL Number: MFCD07369278
CAS Number: 66335-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=166c76d77dfc0cba0882d2f31f9e5ee77376ec1b&vid=166c76d77dfc0cba0882d2f31f9e5ee77376ec1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR