N,N-Dimethyl-N-(methylsulfanylmethylene)ammonium iodide ≥98.0%

Supplier: TCI America

Synonyms: N,N-Dimethyl-N-(methylsulfanylmethylene)iminium Iodide, N,N-Dimethyl-N-(methylsulphanylmethylene)iminium Iodide

D4340-25G D4340-5G
TCD4340-25GEA 481.9 CAD
TCD4340-25G TCD4340-5G
N,N-Dimethyl-N-(methylsulfanylmethylene)ammonium iodide ≥98.0%
N,N-Dimethyl-N-(methylsulfanylmethylene)ammonium iodide

CAS Number: 29085-13-0
MDL Number: MFCD17015287
Molecular Formula: C4H10INS
Molecular Weight: 231.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 125


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: N,N-Dimethyl-N-(methylsulfanylmethylene)iminium Iodide

Formula: C₄H₁₀INS
Melting Pt: 125 °C
MDL Number: MFCD17015287
CAS Number: 29085-13-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=396fd79b04ddcfd351de50835a1329beed3ea688&vid=396fd79b04ddcfd351de50835a1329beed3ea688&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR