2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic dianhydride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D4339-1G
D4339-5G
TCD4339-1GEA
364.85
CAD
TCD4339-1G
TCD4339-5G
2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic dianhydride ≥98.0% (by HPLC, titration analysis)
2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic dianhydride
CAS Number: 83204-68-6
MDL Number: MFCD00406909
Molecular Formula: C14H2Br2O6
Molecular Weight: 425.97
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 350
Formula:
C₁₄H₂Br₂O₆ Melting Pt: 350 °C |
MDL Number:
MFCD00406909 CAS Number: 83204-68-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...