Dibenzo[a,e]cyclooctene ≥97.0%

Supplier: TCI America

Synonyms: DBCOT, Dibenzo[a,e]cyclooctatetraene, Dibenzo[a,e][8]annulene

D4338-1G D4338-200MG
TCD4338-1GEA 770.6 CAD
TCD4338-1G TCD4338-200MG
Dibenzo[a,e]cyclooctene ≥97.0%
Dibenzo[a,e]cyclooctene

CAS Number: 262-89-5
Molecular Formula: C16H12
Molecular Weight: 204.27
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 107
Lambda max.: 304 nm (CH2Cl2)Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: Dibenzo[a,e]cyclooctatetraene, Dibenzo[a,e][8]annulene, DBCOT

Formula: C₁₆H₁₂
MW: 204.27 g/mol
Boiling Pt: 130 °C
Melting Pt: 107 °C
CAS Number: 262-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57894478d2ae189617afdb69a99ec3bf502a0c1a&vid=57894478d2ae189617afdb69a99ec3bf502a0c1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR