(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-N-Despropyl Pramipexole, Pramipexole Related Compound A

D4337-1G D4337-5G
TCD4337-1GEA 107.44 CAD
TCD4337-1G TCD4337-5G
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole ≥98.0% (by GC, titration analysis)
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole

CAS Number: 106092-09-5
MDL Number: MFCD07368003
Molecular Formula: C7H11N3S
Molecular Weight: 169.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 229
Specific rotation [a]20/D: -96 deg (C=1, MeOH)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: (S)-N-Despropyl Pramipexole

Formula: C₇H₁₁N₃S
MW: 169.25 g/mol
Melting Pt: 229 °C
MDL Number: MFCD07368003
CAS Number: 106092-09-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=394584f33a1e81152ad353678d3a0508bf3318b2&vid=394584f33a1e81152ad353678d3a0508bf3318b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR