4-(Difluoromethoxy)benzaldehyde ≥98.0% (by GC)
Supplier: TCI America
D4334-1G
D4334-5G
TCD4334-1GEA
50.56
CAD
TCD4334-1G
TCD4334-5G
4-(Difluoromethoxy)benzaldehyde ≥98.0% (by GC)
4-(Difluoromethoxy)benzaldehyde
CAS Number: 73960-07-3
MDL Number: MFCD00042252
Molecular Formula: C8H6F2O2
Molecular Weight: 172.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Flash Point (°C): 109
Specific Gravity (20/20): 1.31
Lambda max.: 256 nm (EtOH)
Formula:
C₈H₆F₂O₂ Boiling Pt: 87 °C Density: 1.31 (20 °C) Flash Pt: 109 °C |
MDL Number:
MFCD00042252 CAS Number: 73960-07-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...