Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) ≥98.0%

Supplier: TCI America

Synonyms: [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]palladium(II) Dichloride, [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]dichloropalladium(II)

D4333-1G D4333-200MG
TCD4333-1GEA 162.74 CAD
TCD4333-1G TCD4333-200MG
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II) ≥98.0%
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)

CAS Number: 205319-10-4
MDL Number: MFCD14155707
Molecular Formula: C39H32Cl2OP2Pd
Molecular Weight: 755.95
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 287


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction
Other Product Names / Synonyms: [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]dichloropalladium(II), [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]palladium(II) Dichloride

Formula: C₃₉H₃₂Cl₂OP₂Pd
MW: 755.95 g/mol
Melting Pt: 287 °C
MDL Number: MFCD14155707
CAS Number: 205319-10-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc492c74542aac38e2cad46d6811d7988d58095b&vid=bc492c74542aac38e2cad46d6811d7988d58095b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR