4,4-Difluorocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4,4-difluorocyclohexane-1-carboxylic acid

D4330-1G D4330-5G
TCD4330-1GEA 50.56 CAD
TCD4330-1G TCD4330-5G
4,4-Difluorocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
4,4-Difluorocyclohexanecarboxylic acid

CAS Number: 122665-97-8
MDL Number: MFCD03788493
Molecular Formula: C7H10F2O2
Molecular Weight: 164.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 107

Formula: C7H10F2O2
MW: 164.15 g/mol
Melting Pt: 107 °C
MDL Number: MFCD03788493
CAS Number: 122665-97-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3afc8eec224e60ebea4ceefae7d78c734e03413b&vid=3afc8eec224e60ebea4ceefae7d78c734e03413b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR