9,9-Dihexylfluorene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
9,9-Dihexyl-9H-fluorene
D4327-1G
D4327-5G
TCD4327-1GEA
198.29
CAD
TCD4327-1G
TCD4327-5G
9,9-Dihexylfluorene ≥98.0% (by GC)
9,9-Dihexylfluorene
CAS Number: 123863-97-8
MDL Number: MFCD14584648
Molecular Formula: C25H34
Molecular Weight: 334.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 32
Formula:
C₂₅H₃₄ MW: 334.55 g/mol Melting Pt: 32 °C |
MDL Number:
MFCD14584648 CAS Number: 123863-97-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...