3,5-Dibromopyridine-N-oxide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3,5-Dibromopyridine1-oxide

D4326-1G D4326-5G
TCD4326-1GEA 97.49 CAD
TCD4326-1G TCD4326-5G
3,5-Dibromopyridine-N-oxide ≥98.0% (by GC)
3,5-Dibromopyridine N-oxide

CAS Number: 2402-99-5
MDL Number: MFCD00160750
Molecular Formula: C5H3Br2NO
Molecular Weight: 252.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 144

Formula: C₅H₃Br₂NO
MW: 252.8914 g/mol
Melting Pt: 144 °C
MDL Number: MFCD00160750
CAS Number: 2402-99-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a66b3d32b9e8b05cd59ccfa1b0c05944b0f6900&vid=7a66b3d32b9e8b05cd59ccfa1b0c05944b0f6900&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR