2,5-Dibromopyrazine ≥98.0% (by GC)

Supplier: TCI America

D4323-1G D4323-5G
TCD4323-1GEA 39.5 CAD
TCD4323-1G TCD4323-5G
2,5-Dibromopyrazine ≥98.0% (by GC)
2,5-Dibromopyrazine

CAS Number: 23229-26-7
MDL Number: MFCD08061601
Molecular Formula: C4H2Br2N2
Molecular Weight: 237.88
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 47
Storage Temperature: 0-10°C

Formula: C₄H₂Br₂N₂
Melting Pt: 47 °C
MDL Number: MFCD08061601
CAS Number: 23229-26-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba3dbc1b840bc4a43a1f3d01044faf521773ef&vid=d0ba3dbc1b840bc4a43a1f3d01044faf521773ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR