(2,4-Difluorophenyl)acetic acid ≥98.0%
Supplier: TCI America
Synonyms:
2,4-Difluorophenylacetic acid
D4321-25G
D4321-5G
TCD4321-25GEA
584.6
CAD
TCD4321-25G
TCD4321-5G
(2,4-Difluorophenyl)acetic acid ≥98.0%
(2,4-Difluorophenyl)acetic acid
CAS Number: 81228-09-3
MDL Number: MFCD00009999
Molecular Formula: C8H6F2O2
Molecular Weight: 172.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 117
Formula:
(C₆H₃F₂)CH₂COOH MW: 172.13 g/mol Boiling Pt: 259 Melting Pt: 117 °C |
MDL Number:
MFCD00009999 CAS Number: 81228-09-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...