3,3-Difluoropyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3,3-Difluoropyrrolidine HCl

D4315-1G D4315-200MG
TCD4315-1GEA 231.47 CAD
TCD4315-1G TCD4315-200MG
3,3-Difluoropyrrolidine hydrochloride ≥98.0% (by titrimetric analysis)
3,3-Difluoropyrrolidine hydrochloride

CAS Number: 163457-23-6
MDL Number: MFCD03788948
Molecular Formula: C4H7F2N
Molecular Weight: 143.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 134

Formula: C₄H₇F₂N
MW: 143.56 g/mol
Melting Pt: 134 °C
MDL Number: MFCD03788948
CAS Number: 163457-23-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f3c906d63a7077cb33f8c936ee59ebc46069f45&vid=7f3c906d63a7077cb33f8c936ee59ebc46069f45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR