Diphenylbis(phenylethynyl)silane ≥98.0% (by GC)

Supplier: TCI America

D4312-1G D4312-5G
TCD4312-1GEA 94.17 CAD
TCD4312-1G TCD4312-5G
Diphenylbis(phenylethynyl)silane ≥98.0% (by GC)
Diphenylbis(phenylethynyl)silane

CAS Number: 18784-61-7
MDL Number: MFCD00093508
Molecular Formula: C28H20Si
Molecular Weight: 384.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 81

Formula: C₂₈H₂₀Si
MW: 384.55 g/mol
Melting Pt: 81 °C
MDL Number: MFCD00093508
CAS Number: 18784-61-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e15f16e41ba43c548e4c8a73f1dfcefcacd34b1&vid=5e15f16e41ba43c548e4c8a73f1dfcefcacd34b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR