Di-tert-butyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Di-tert-butyl Ester
D4311-1G
D4311-5G
TCD4311-1GEA
144.65
CAD
TCD4311-1G
TCD4311-5G
Di-tert-butyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥98.0% (by HPLC, titration analysis)
Di-tert-butyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
CAS Number: 55536-71-5
MDL Number: MFCD01882057
Molecular Formula: C17H27NO4
Molecular Weight: 309.41
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 156
Product Categories: Synthetic Organic Chemistry, Reduction
Other Product Names / Synonyms: 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Di-tert-butyl Ester
Formula:
C₁₇H₂₇NO₄ MW: 309.41 g/mol Melting Pt: 156 °C |
MDL Number:
MFCD01882057 CAS Number: 55536-71-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...