9,9-Dimethyl-9H-9-silafluorene ≥98.0% (by GC)

Supplier: TCI America

D4308-1G D4308-5G
TCD4308-1GEA 240.16 CAD
TCD4308-1G TCD4308-5G
9,9-Dimethyl-9H-9-silafluorene ≥98.0% (by GC)
9,9-Dimethyl-9H-9-silafluorene

CAS Number: 13688-68-1
Molecular Formula: C14H14Si
Molecular Weight: 210.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 60


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 5,5-Dimethyl-5H-dibenzo[b,d]silole

Formula: C₁₄H₁₄Si
Melting Pt: 60 °C
MDL Number: MFCD00093801
CAS Number: 13688-68-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e8a66fe2709db10bec3f9dcba6b09ac1fc6de2f&vid=2e8a66fe2709db10bec3f9dcba6b09ac1fc6de2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR