2,5-Dibromo-3-methylthiophene ≥98.0% (by GC)

Supplier: TCI America

D4304-25G D4304-5G
TCD4304-25GEA 270.18 CAD
TCD4304-25G TCD4304-5G
2,5-Dibromo-3-methylthiophene ≥98.0% (by GC)
2,5-Dibromo-3-methylthiophene

CAS Number: 13191-36-1
MDL Number: MFCD00015470
Molecular Formula: C5H4Br2S
Molecular Weight: 255.96
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 110
Specific Gravity (20/20): 1.99
Storage Temperature: 0-10°C

Formula: C₅H₄Br₂S
Boiling Pt: 108 °C
Density: 1.99 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00015470
CAS Number: 13191-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9454d2e9643134aee377d9a0362e352fa449c99e&vid=9454d2e9643134aee377d9a0362e352fa449c99e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR