Doxofylline ≥98.0%
Supplier: TCI America
Synonyms:
7-(1,3-Dioxolan-2-ylmethyl)theophylline, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethylxanthine, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethyl-1H-purine-2,6-dione
CAS Number: 69975-86-6
MDL Number: MFCD00865218
Molecular Formula: C11H14N4O4
Molecular Weight: 266.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 144
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients, Nucleobases and Their Analogs, Purine Alkaloids
Other Product Names / Synonyms: 7-(1,3-Dioxolan-2-ylmethyl)theophylline, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethyl-1H-purine-2,6-dione, 7-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethylxanthine
Formula:
C₁₁H₁₄N₄O₄ MW: 266.26 g/mol Melting Pt: 144 °C |
MDL Number:
MFCD00865218 CAS Number: 69975-86-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...