Diisopropyl trisulfide ≥98.0% (by GC)

Supplier: TCI America

D4300-25G D4300-5G
TCD4300-25GEA 570.38 CAD
TCD4300-25G TCD4300-5G
Diisopropyl trisulfide ≥98.0% (by GC)
Diisopropyl trisulfide

CAS Number: 5943-34-0
MDL Number: MFCD00040042
Molecular Formula: C6H14S3
Molecular Weight: 182.36
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Specific Gravity (20/20): 1.04

Formula: C₆H₁₄S₃
Boiling Pt: 79 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00040042
CAS Number: 5943-34-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47f9638cfa566c1085724abbc12b298835feba3f&vid=47f9638cfa566c1085724abbc12b298835feba3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR