1,11-Dioxa[11]paracyclophane ≥97.0% (by GC)
Supplier: TCI America
D4298-1G
D4298-250MG
TCD4298-1GEA
471.16
CAD
TCD4298-1G
TCD4298-250MG
1,11-Dioxa[11]paracyclophane ≥97.0% (by GC)
1,11-Dioxa[11]paracyclophane
CAS Number: 6571-51-3
Molecular Formula: C15H22O2
Molecular Weight: 234.34
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 55
Formula:
C₁₅H₂₂O₂ Melting Pt: 55 °C |
CAS Number:
6571-51-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...