5,6-Dichlorobenzimidazole Hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D4295-1G
D4295-5G
TCD4295-1GEA
171.9
CAD
TCD4295-1G
TCD4295-5G
5,6-Dichlorobenzimidazole Hydrochloride ≥98.0% (by HPLC, titration analysis)
5,6-Dichlorobenzimidazole Hydrochloride
CAS Number: 1087737-96-9
MDL Number: MFCD16810292
Molecular Formula: C7H4Cl2N2
Molecular Weight: 223.48
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Formula:
C₇H₄Cl₂N₂ MW: 223.48 g/mol |
MDL Number:
MFCD16810292 CAS Number: 1087737-96-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...