1,3-Dibromo-2-iodobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,6-Dibromoiodobenzene

D4294-1G D4294-5G
TCD4294-1GEA 110.6 CAD
TCD4294-1G 76219-032
1,3-Dibromo-2-iodobenzene ≥98.0% (by GC)
1,3-Dibromo-2-iodobenzene

CAS Number: 19821-80-8
MDL Number: MFCD00192666
Molecular Formula: C6H3Br2I
Molecular Weight: 361.80
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 100

Formula: C₆H₃Br₂I
MW: 361.80 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00192666
CAS Number: 19821-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b13569194b4bf5b89fa8fa20da4d285ca5d917c0&vid=b13569194b4bf5b89fa8fa20da4d285ca5d917c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR