2,3-Dimethylbenzonitrile ≥97.0% (by GC)

Supplier: TCI America

D4293-25G D4293-5G
TCD4293-25GEA 401.64 CAD
TCD4293-25G TCD4293-5G
2,3-Dimethylbenzonitrile ≥97.0% (by GC)
2,3-Dimethylbenzonitrile

CAS Number: 5724-56-1
MDL Number: MFCD00019744
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Melting point (°C): 16
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 3-Cyano-o-xylene

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 62 °C
Melting Pt: 16 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00019744
CAS Number: 5724-56-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bc8f55812f761c249be62ae24c713a5c2f45f5f&vid=0bc8f55812f761c249be62ae24c713a5c2f45f5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR