5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside ≥98.0%

Supplier: TCI America

Synonyms: 5,6-Dichloro-1-(β-D-ribofuranosyl)benzimidazole, 5,6-Dichlorobenzimidazole Riboside, (2R,3R,4S,5R)-2-(5,6-Dichloro-1H-benzo[d]imidazol-1-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, DRB

D4292-100MG D4292-1G
TCD4292-100MGEA 277.23 CAD
TCD4292-100MG TCD4292-1G
5,6-Dichlorobenzimidazole-1-β-D-Ribofuranoside ≥98.0%
5,6-Dichlorobenzimidazole-1-beta-D-Ribofuranoside

CAS Number: 53-85-0
MDL Number: MFCD00036785
Molecular Formula: C12H12Cl2N2O4
Molecular Weight: 319.14
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 222
Specific rotation [a]20/D: -67 deg (C=2, Pyridine)
Lambda max.: 296 nmStorage Temperature: <0°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 5,6-Dichlorobenzimidazole Riboside, 5,6-Dichloro-1-(beta-D-ribofuranosyl)benzimidazole, DRB

Formula: C₁₂H₁₂Cl₂N₂O₄
MW: 319.14 g/mol
Melting Pt: 222 °C
MDL Number: MFCD00036785
CAS Number: 53-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=872e81efeea0016ee61923d743236de631b089da&vid=872e81efeea0016ee61923d743236de631b089da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR