4-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-(4-Dodecyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Dodecyl-2-thiopheneboronic Acid Pinacol Ester
CAS Number: 1173788-58-3
Molecular Formula: C22H39BO2S
Molecular Weight: 378.42
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.96
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-(4-Dodecyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Dodecyl-2-thiopheneboronic Acid Pinacol Ester
Formula:
C₂₂H₃₉BO₂S MW: 378.42 g/mol Density: 0.96 (20 °C) |
CAS Number:
1173788-58-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...