3-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥95.0% (by GC)

Supplier: TCI America

D4290-1G D4290-5G
TCD4290-1GEA 233.84 CAD
TCD4290-1G TCD4290-5G
3-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥95.0% (by GC)
3-Dodecyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene

CAS Number: 960524-18-9
Molecular Formula: C22H39BO2S
Molecular Weight: 378.42
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.95
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-(3-Dodecyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Dodecyl-2-thiopheneboronic Acid Pinacol Ester

Formula: C₂₂H₃₉BO₂S
Density: 0.95 (20 °C)
CAS Number: 960524-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22a9ab0c8b629bebc07edf24a69f5c8e15995e09&vid=22a9ab0c8b629bebc07edf24a69f5c8e15995e09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR