2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D4289-25G D4289-5G
TCD4289-25GEA 882.59 CAD
TCD4289-25G TCD4289-5G
2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulfonyl)imide ≥98.0% (by titrimetric analysis)
2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulphonyl)imide

CAS Number: 169051-76-7
MDL Number: MFCD03788922
Molecular Formula: C10H15F6N3O4S2
Molecular Weight: 419.36
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Melting point (°C): 8
Specific Gravity (20/20): 1.46


Product Categories: Synthetic Organic Chemistry, Ionic Liquids
Other Product Names / Synonyms: 1,2-Dimethyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide

Formula: C₁₀H₁₅F₆N₃O₄S₂
Melting Pt: 8 °C
Density: 1.46 (20 °C)
MDL Number: MFCD03788922
CAS Number: 169051-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da30932aea86a78efe7b994f6f448dd62738d202&vid=da30932aea86a78efe7b994f6f448dd62738d202&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR