2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine ≥98.0%

Supplier: TCI America

Synonyms: 2,6-Dichloro-7-deazapurine

D4284-1G D4284-5G
TCD4284-1GEA 53.72 CAD
TCD4284-1G TCD4284-5G
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine ≥98.0%
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine

CAS Number: 90213-66-4
MDL Number: MFCD08059278
Molecular Formula: C6H3Cl2N3
Molecular Weight: 188.01
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 251


Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine

Formula: C₆H₃Cl₂N₃
Melting Pt: 251 °C
MDL Number: MFCD08059278
CAS Number: 90213-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b106ec9f583bbc34f098b3f72fe9cf7cba3ceb&vid=c1b106ec9f583bbc34f098b3f72fe9cf7cba3ceb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR