Dipropyleneglycol dimethyl ether (mixture of isomers) ≥94.0% (by GC)

Supplier: TCI America

D4283-25ML D4283-500ML
TCD4283-25MLEA 36.34 CAD
TCD4283-25ML TCD4283-500ML
Dipropyleneglycol dimethyl ether (mixture of isomers) ≥94.0% (by GC)
Dipropyleneglycol dimethyl ether (mixture of isomers)

CAS Number: 111109-77-4
MDL Number: MFCD00210047
Molecular Formula: C8H18O3
Molecular Weight: 162.23
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Flash Point (°C): 65
Specific Gravity (20/20): 0.90

Formula: C₈H₁₈O₃
Density: 0.90 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00210047
CAS Number: 111109-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f2daddb0dcbf114052d47672153eaf6ffdbf811&vid=7f2daddb0dcbf114052d47672153eaf6ffdbf811&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR