2,4-Dibromobenzoic acid ≥98.0%

Supplier: TCI America

D4278-1G D4278-5G
TCD4278-1GEA 134.3 CAD
TCD4278-1G TCD4278-5G
2,4-Dibromobenzoic acid ≥98.0%
2,4-Dibromobenzoic acid

CAS Number: 611-00-7
MDL Number: MFCD00234253
Molecular Formula: C7H4Br2O2
Molecular Weight: 279.92
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173

Formula: C₇H₄Br₂O₂
MW: 279.92
Melting Pt: 173 °C
MDL Number: MFCD00234253
CAS Number: 611-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04d6e2b7f4b3ae6001bb2de42c8ecb39a3b7cdea&vid=04d6e2b7f4b3ae6001bb2de42c8ecb39a3b7cdea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR