2,2-Dibromomalonamide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
D4272-25G
D4272-5G
TCD4272-25GEA
165.9
CAD
TCD4272-25G
TCD4272-5G
2,2-Dibromomalonamide ≥98.0% (by total nitrogen basis)
2,2-Dibromomalonamide
CAS Number: 73003-80-2
MDL Number: MFCD00014803
Molecular Formula: C3H4Br2N2O2
Molecular Weight: 259.89
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 204
Other Product Names / Synonyms: Dibromomalonic Diamide
Formula:
C₃H₄Br₂N₂O₂ Melting Pt: 204 °C |
MDL Number:
MFCD00014803 CAS Number: 73003-80-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...