2,5-Dichloro-3-nitropyridine ≥98.0% (by GC)

Supplier: TCI America

D4267-25G D4267-5G
TCD4267-25GEA 390.1 CAD
TCD4267-25G TCD4267-5G
2,5-Dichloro-3-nitropyridine ≥98.0% (by GC)
2,5-Dichloro-3-nitropyridine

CAS Number: 21427-62-3
MDL Number: MFCD06658963
Molecular Formula: C5H2Cl2N2O2
Molecular Weight: 192.98
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 45
Flash Point (°C): 110

Formula: C₅H₂Cl₂N₂O₂
Melting Pt: 45 °C
Flash Pt: 110 °C
MDL Number: MFCD06658963
CAS Number: 21427-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bb52140fb93d6d1651c049bfd6a1402b1691142&vid=3bb52140fb93d6d1651c049bfd6a1402b1691142&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR