[(2,6-Dimethylphenyl)amino](oxo)acetic Acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D4265-1G D4265-5G
TCD4265-1GEA 161.16 CAD
TCD4265-1G TCD4265-5G
[(2,6-Dimethylphenyl)amino](oxo)acetic Acid ≥98.0% (by HPLC, titration analysis)
[(2,6-Dimethylphenyl)amino](oxo)acetic Acid

CAS Number: 2903-48-2
MDL Number: MFCD10018453
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: (2,6-Dimethylanilino)(oxo)acetic Acid, N-(2,6-Dimethylphenyl)oxamic Acid, DMPAO

Formula: C₁₀H₁₁NO₃
Melting Pt: 173 °C
MDL Number: MFCD10018453
CAS Number: 2903-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2bc9ce98f9414e6928b9eb7918a4c630b098c37&vid=b2bc9ce98f9414e6928b9eb7918a4c630b098c37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR