2-(Diisopropylsilyl)pyridine ≥95.0% (by GC)

Supplier: TCI America

D4264-1G D4264-5G
TCD4264-1GEA 122.13 CAD
TCD4264-1G TCD4264-5G
2-(Diisopropylsilyl)pyridine ≥95.0% (by GC)
2-(Diisopropylsilyl)pyridine

CAS Number: 1232692-92-0
MDL Number: MFCD22200515
Molecular Formula: C11H19NSi
Molecular Weight: 193.37
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 84
Specific Gravity (20/20): 0.90


Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, C-H Bond Activation Reaction
Other Product Names / Synonyms: Diisopropyl(2-pyridyl)silane, PyDipSiH

Formula: C₁₁H₁₉NSi
Density: 0.90 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD22200515
CAS Number: 1232692-92-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1de31f84078b32a28a516a1f09d92024641ae91d&vid=1de31f84078b32a28a516a1f09d92024641ae91d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR