2,4-Dibutoxyphenylboronic acid 97-108% (contains varying amounts of Anhydride)
Supplier: TCI America
D4263-1G
D4263-5G
TCD4263-1GEA
537.2
CAD
TCD4263-1G
TCD4263-5G
2,4-Dibutoxyphenylboronic acid 97-108% (contains varying amounts of Anhydride)
2,4-Dibutoxyphenylboronic acid
CAS Number: 870778-89-5
MDL Number: MFCD07784348
Molecular Formula: C14H23BO4
Molecular Weight: 266.14
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 104
Other Product Names / Synonyms: 2,4-Dibutoxybenzeneboronic Acid
Formula:
C₁₄H₂₃BO₄ Melting Pt: 104 °C |
MDL Number:
MFCD07784348 CAS Number: 870778-89-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...