1-(2,6-Dichlorophenyl)oxindole ≥98.0% (by GC)
Supplier: TCI America
D4260-25G
D4260-5G
TCD4260-25GEA
233.05
CAD
TCD4260-25G
TCD4260-5G
1-(2,6-Dichlorophenyl)oxindole ≥98.0% (by GC)
1-(2,6-Dichlorophenyl)oxindole
CAS Number: 15362-40-0
MDL Number: MFCD00451393
Molecular Formula: C14H9Cl2NO
Molecular Weight: 278.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 127
Other Product Names / Synonyms: 1-(2,6-Dichlorophenyl)-2-oxoindoline, 1-(2,6-Dichlorophenyl)-2-indolinone, Diclofenac Amide
Formula:
C₁₄H₉Cl₂NO Melting Pt: 127 °C |
MDL Number:
MFCD00451393 CAS Number: 15362-40-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...