N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide ≥98.0%

Supplier: TCI America

Synonyms: N-(3-(3-(Dimethylamino)acryloyl)phenyl)-N-ethylacetamide

D4259-1G D4259-5G
TCD4259-1GEA 45.82 CAD
TCD4259-1G TCD4259-5G
N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide ≥98.0%
N-[3-[3-(Dimethylamino)acryloyl]phenyl]-N-ethylacetamide

CAS Number: 96605-66-2
MDL Number: MFCD04117958
Molecular Formula: C15H20N2O2
Molecular Weight: 260.34
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 113

Formula: C₁₅H₂₀N₂O₂
MW: 260.34 g/mol
Melting Pt: 113 °C
MDL Number: MFCD04117958
CAS Number: 96605-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34c9086bb66fa80d68d987d02320bb8b854e5b5a&vid=34c9086bb66fa80d68d987d02320bb8b854e5b5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR