N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-10, SB-10) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier: TCI America
Synonyms:
3-(Decyldimethylazaniumyl)propane-1-sulfonate, 3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulphonate, 3-(Decyldimethylazaniumyl)propane-1-sulphonate, 3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulfonate
[for Biochemical Research]
CAS Number: 15163-36-7
MDL Number: MFCD00036908
Molecular Formula: C15H33NO3S
Molecular Weight: 307.49
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 229
Product Categories: Bioscience, Amphoteric Surfactants (for Biochemistry)
Other Product Names / Synonyms: 3-(Decyldimethylammonio)propane-1-sulfonate, Caprylyl Sulfobetaine
Formula:
C₁₅H₃₃NO₃S MW: 307.4924 g/mol Melting Pt: 229 °C |
MDL Number:
MFCD00036908 CAS Number: 15163-36-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...