1,1-Dimethyl-1,3-dihydrobenzo[c][1,2]oxasilole ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dihydro-1,1-dimethyl-2,1-benzoxasilole

D4243-1G D4243-5G
TCD4243-1GEA 237 CAD
TCD4243-1G TCD4243-5G
1,1-Dimethyl-1,3-dihydrobenzo[c][1,2]oxasilole ≥98.0%
1,1-Dimethyl-1,3-dihydrobenzo[c][1,2]oxasilole

CAS Number: 321903-29-1
MDL Number: MFCD20922818
Molecular Formula: C9H12OSi
Molecular Weight: 164.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 45
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 1,3-Dihydro-1,1-dimethyl-2,1-benzoxasilole

Formula: C₉H₁₂OSi
MW: 164.28 g/mol
Boiling Pt: 45 °C
Density: 1.02 (20 °C)
MDL Number: MFCD20922818
CAS Number: 321903-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80a1e253a9036985b078e127548e76a2491e8fac&vid=80a1e253a9036985b078e127548e76a2491e8fac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR