2-(Dimethylamino)-2-oxoacetic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D4241-5G D4241-1G
TCD4241-5GEA 132.72 CAD
TCD4241-5G TCD4241-1G
2-(Dimethylamino)-2-oxoacetic acid ≥97.0% (by titrimetric analysis)
2-(Dimethylamino)-2-oxoacetic acid

CAS Number: 32833-96-8
MDL Number: MFCD01971023
Molecular Formula: C4H7NO3
Molecular Weight: 117.10
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 125


Other Product Names / Synonyms: N,N-Dimethyloxamidic Acid

Formula: C₄H₇NO₃
Melting Pt: 125 °C
MDL Number: MFCD01971023
CAS Number: 32833-96-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8b882faba4de494791cadad1f07ca6474bbade9&vid=e8b882faba4de494791cadad1f07ca6474bbade9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR