4,4'-Diethynylbiphenyl ≥98.0%
Supplier: TCI America
Synonyms:
4,4'-Diethynyl-1,1'-biphenyl
D4233-1G
D4233-200MG
TCD4233-1GEA
704.21
CAD
TCD4233-1G
TCD4233-200MG
4,4'-Diethynylbiphenyl ≥98.0%
4,4'-Diethynylbiphenyl
CAS Number: 38215-38-2
MDL Number: MFCD00458964
Molecular Formula: C16H10
Molecular Weight: 202.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 168
Formula:
C₁₆H₁₀ Melting Pt: 168 °C |
MDL Number:
MFCD00458964 CAS Number: 38215-38-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...